Assay Details
Gene Symbol: RGD1309170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134579.1 | NP_001128051.1 | 4 - 5 | 862 | 98 | |
XM_006233387.2 | XP_006233449.1 | 5 - 6 | 758 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
