Assay Details
| Gene Symbol: RGD1309170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134579.1 | NP_001128051.1 | 5 - 6 | 947 | 97 | |
| XM_006233387.2 | XP_006233449.1 | 6 - 7 | 843 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

RGD1309170
Genomic Map