Assay Details
| Gene Symbol: NEWGENE_1305281 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233223.2 | XP_006233285.1 | 4 - 5 | 635 | 150 | |
| XM_006233224.2 | XP_006233286.1 | 4 - 5 | 750 | 150 | ||
| XM_008761473.1 | XP_008759695.1 | 4 - 5 | 506 | 150 | ||
| Gene Symbol: Hiat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106467.3 | NP_001099937.1 | 503 | 150 | ||
| GenBank mRNA | BC160912.1 | - | 492 | 7456962 | 150 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

NEWGENE_1305281
Genomic Map