Assay Details
Gene Symbol: Hiat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106467.3 | NP_001099937.1 | 616 | 145 | ||
GenBank mRNA | BC160912.1 | - | 605 | 7456962 | 145 |
Gene Symbol: NEWGENE_1305281 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233223.2 | XP_006233285.1 | 5 - 6 | 748 | 145 | |
XM_006233224.2 | XP_006233286.1 | 5 - 6 | 863 | 145 | ||
XM_008761473.1 | XP_008759695.1 | 5 - 6 | 619 | 145 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
