Assay Details
Gene Symbol: Ahcyl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108561.1 | NP_001102031.1 | 2 - 3 | 225 | 68 | |
XM_006233165.1 | XP_006233227.1 | 1 - 2 | 109 | 68 | ||
XM_006233166.2 | XP_006233228.1 | 2 - 3 | 449 | 68 | ||
XM_006233167.1 | XP_006233229.1 | 1 - 2 | 109 | 68 | ||
XM_008761405.1 | XP_008759627.1 | 2 - 3 | 461 | 68 | ||
XM_008761406.1 | XP_008759628.1 | 2 - 3 | 461 | 68 | ||
GenBank mRNA | BC169126.1 | - | 1 - 2 | 89 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
