Assay Details
Gene Symbol: Kcna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233132.2 | XP_006233194.1 | 3 | 11152 | 64 | |
XM_006233133.2 | XP_006233195.1 | 3 | 10734 | 64 | ||
XM_006233134.2 | XP_006233196.1 | 3 | 10717 | 64 | ||
XM_006233135.2 | XP_006233197.1 | 3 | 10521 | 64 | ||
XM_008761371.1 | XP_008759593.1 | 3 | 10762 | 64 | ||
GenBank mRNA | FQ213529.1 | - | 1 | 1381 | 64 | |
FQ233516.1 | - | 1 | 1132 | 64 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
