Assay Details
| Gene Symbol: Aknad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044303.2 | NP_001037768.2 | 11 - 12 | 2016 | 81 | |
| XM_008761422.1 | XP_008759644.1 | 11 - 12 | 3197 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aknad1
Genomic Map