Assay Details
| Gene Symbol: LOC100910501 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008761484.1 | XP_008759706.1 | 1 - 2 | 885 | 89 | |
| Gene Symbol: Enpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022251.2 | NP_071587.2 | 1 - 2 | 1037 | 89 | |
| GenBank mRNA | AF214568.1 | - | 1 - 2 | 854 | 89 | |
| BC066663.1 | - | 1 - 2 | 859 | 5623429 | 89 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100910501
Genomic Map