Assay Details
| Gene Symbol: Acadm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016986.2 | NP_058682.2 | 3 - 4 | 323 | 66 | |
| GenBank mRNA | CK359511.1 | - | 3 - 4 | 238 | 66 | |
| J02791.1 | - | 3 - 4 | 238 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acadm
Genomic Map