Assay Details
| Gene Symbol: Dnajb14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109193.2 | NP_001102663.2 | 5 - 6 | 761 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajb14| Gene Symbol: Dnajb14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109193.2 | NP_001102663.2 | 5 - 6 | 761 | 68 | |
Set Membership: |
|