Assay Details
| Gene Symbol: Slc44a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013914.1 | NP_001013936.1 | 13 - 14 | 1872 | 68 | |
| GenBank mRNA | BC079142.1 | - | 13 - 14 | 1872 | 7128044 | 68 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P154859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165742 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392192 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648584 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409853 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673077 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107093 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125806 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P154859 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P154859 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P154859 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 21024 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 73125 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91265 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99464 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 47029 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55016 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10826463 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826462 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826460 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826459 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826454 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826465 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826467 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826466 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826452 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826453 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826458 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826468 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826461 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826455 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826464 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10826451 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200031871.rn.1 | TC0200004579.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200031872.rn.1 | TC0200004579.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200031870.rn.1 | TC0200004579.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10826451 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20921275 | - | - |

Slc44a3
Genomic Map