Assay Details
| Gene Symbol: Rab5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC072495.1 | - | 215 | 81 | ||
| BC161848.1 | - | 230 | 9025210 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC072495.1, BC161848.1 transcripts.
Set Membership: |
|

Rab5a
Genomic Map