Assay Details
| Gene Symbol: Rab5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022692.1 | NP_073183.1 | 162 | 143 | ||
| GenBank mRNA | AF072935.1 | - | 162 | 143 | ||
| BC161848.1 | - | 488 | 9025210 | 143 | ||
| Gene Symbol: Rab5al1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751377.3 | XP_003751425.1 | 1 | 394 | 143 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rab5a
Genomic Map