Assay Details
Gene Symbol: Mbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011924.1 | NP_001011924.1 | 2 - 3 | 258 | 67 | |
XM_006254909.2 | XP_006254971.1 | 2 - 3 | 403 | 67 | ||
XM_006254910.2 | XP_006254972.1 | 2 - 3 | 403 | 67 | ||
XM_006254911.2 | XP_006254973.1 | 2 - 3 | 403 | 67 | ||
XM_006254913.2 | XP_006254975.1 | 2 - 3 | 402 | 67 | ||
XM_006254914.2 | XP_006254976.1 | 2 - 3 | 402 | 67 | ||
XM_008772105.1 | XP_008770327.1 | 2 - 3 | 405 | 67 | ||
GenBank mRNA | BC081932.1 | - | 2 - 3 | 258 | 7114194 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
