Assay Details
Gene Symbol: Mrpl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106205.2 | NP_001099675.1 | 1 - 2 | 145 | 132 | |
GenBank mRNA | BC167033.1 | - | 1 - 2 | 145 | 8372474 | 132 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC167033.1,NM_001106205.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
