Assay Details
Gene Symbol: T2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161835.1 | NP_001155307.1 | 9 - 10 | 965 | 62 | |
XM_006227933.2 | XP_006227995.1 | 9 - 10 | 1927 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
