Assay Details
| Gene Symbol: LOC683722 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101008.1 | NP_001094478.1 | 10 - 11 | 1074 | 110 | |
| XM_008758772.1 | XP_008756994.1 | 10 - 11 | 1485 | 110 | ||
| XM_008758773.1 | XP_008756995.1 | 9 - 10 | 1422 | 110 | ||
| XM_008758774.1 | XP_008756996.1 | 9 - 10 | 1119 | 110 | ||
| GenBank mRNA | BC098026.1 | - | 10 - 11 | 1074 | 7455942 | 110 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC683722
Genomic Map