Assay Details
| Gene Symbol: Zfp455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173314.2 | NP_775436.2 | 2 - 3 | 204 | 91 | |
| XM_006232101.2 | XP_006232163.1 | 2 - 3 | 440 | 91 | ||
| GenBank mRNA | BC161833.1 | - | 2 - 3 | 204 | 91 | |
| U67083.1 | - | 192 | 91 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp455
Genomic Map