Assay Details
| Gene Symbol: Hmgb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173341.1 | NP_001166812.1 | 4 - 5 | 532 | 79 | |
| XM_006251014.2 | XP_006251076.1 | 4 - 5 | 620 | 79 | ||
| GenBank mRNA | BC158739.1 | - | 3 - 4 | 472 | 9099241 | 79 |
| EV772004.1 | - | 4 - 5 | 553 | 79 | ||
| Gene Symbol: LOC100912369 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751682.3 | XP_003751730.1 | 4 - 5 | 620 | 79 | |
| XM_006253886.2 | XP_006253948.1 | 4 - 5 | 561 | 79 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hmgb3
Genomic Map