Assay Details
Gene Symbol: Bin2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009524.1 | NP_001009524.1 | 11 - 12 | 1291 | 73 | |
XM_006242737.2 | XP_006242799.1 | 11 - 12 | 1169 | 73 | ||
GenBank mRNA | AF299298.1 | - | 11 - 12 | 1145 | 73 | |
AF414431.1 | - | 11 - 12 | 1145 | 73 | ||
AY040870.1 | - | 11 - 12 | 1291 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
