Assay Details
| Gene Symbol: Npsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106808.1 | NP_001100278.1 | 8 - 9 | 926 | 96 | |
| XM_008766049.1 | XP_008764271.1 | 7 - 8 | 1169 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P004521 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P004516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325872 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188921 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189569 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629318 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513106 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318135 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951374 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181957 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775459 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710949 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85820 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592799 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450923 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958182 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568820 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434029 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15652 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528531 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30655 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121628 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151645 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19554 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P004516 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P004521 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P004521 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P004516 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P004521 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P004516 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P260615 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49093 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1867 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 21111 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14877 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 89579 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57682 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10908669 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908664 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908668 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908667 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908663 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908665 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908661 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908666 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908662 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10908660 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800001799.rn.1 | TC0800000270.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800001798.rn.1 | TC0800000270.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10908660 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22419652 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1360449 | - | - |

Npsr1
Genomic Map