Assay Details
| Gene Symbol: Zscan12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002728485.4 | XP_002728531.1 | 1 - 2 | 1219 | 83 | |
| GenBank mRNA | BC161917.1 | - | 1 - 2 | 934 | 83 | |
More Information
Set Membership: |
|

Zscan12
Genomic Map