Assay Details
| Gene Symbol: Vbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006229645.2 | XP_006229707.1 | 4 - 5 | 555 | 80 | |
| XM_006229646.2 | XP_006229708.1 | 4 - 5 | 570 | 80 | ||
More Information
Set Membership: |
|

Vbp1
Genomic Map