Assay Details
| Gene Symbol: Ddo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109465.2 | NP_001102935.1 | 1 - 2 | 61 | 73 | |
| Gene Symbol: LOC100911156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751978.3 | XP_003752026.1 | 1 - 2 | 123 | 73 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109465.2,XM_003751978.3 transcripts.
Set Membership: |
|

Ddo
Genomic Map