Assay Details
Gene Symbol: Sapcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004069.2 | NP_001004069.1 | 1 - 2 | 121 | 98 | |
XM_006256068.2 | XP_006256130.1 | 1 - 2 | 272 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
