Assay Details
Gene Symbol: Gpsm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003974.2 | NP_001003974.1 | 1 - 2 | 343 | 91 | |
GenBank mRNA | BC088220.1 | - | 1 - 2 | 343 | 7384543 | 91 |
FQ229559.1 | - | 1 - 2 | 163 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC088220.1, NM_001003974.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
