Assay Details
Gene Symbol: Cyp21a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057101.2 | NP_476442.2 | 3 - 4 | 447 | 113 | |
XM_006255907.2 | XP_006255969.1 | 3 - 4 | 524 | 113 | ||
GenBank mRNA | U56853.1 | - | 3 - 4 | 448 | 113 | |
U56854.1 | - | 2 - 3 | 512 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
