Assay Details
| Gene Symbol: Clic1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002807.2 | NP_001002807.1 | 1 - 2 | 274 | 99 | |
| XM_008772745.1 | XP_008770967.1 | 1 - 2 | 275 | 99 | ||
| GenBank mRNA | BC099823.1 | - | 1 - 2 | 226 | 7932557 | 99 |
| EU148064.1 | - | 1 - 2 | 37 | 99 | ||
| FM067949.1 | - | 1 - 2 | 274 | 99 | ||
| FQ219968.1 | - | 1 - 2 | 264 | 99 | ||
| FQ227136.1 | - | 1 - 2 | 60 | 99 | ||
| FQ227567.1 | - | 1 - 2 | 51 | 99 | ||
| FQ227951.1 | - | 1 - 2 | 51 | 99 | ||
| FQ233892.1 | - | 1 - 2 | 51 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the EU148064.1 transcripts.
Set Membership: |
|

Clic1
Genomic Map