Assay Details
Gene Symbol: Clic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002807.2 | NP_001002807.1 | 2 - 3 | 373 | 66 | |
XM_008772745.1 | XP_008770967.1 | 2 - 3 | 374 | 66 | ||
GenBank mRNA | BC099823.1 | - | 2 - 3 | 325 | 7932557 | 66 |
EU148064.1 | - | 2 - 3 | 136 | 66 | ||
FM067949.1 | - | 2 - 3 | 373 | 66 | ||
FQ219968.1 | - | 2 - 3 | 363 | 66 | ||
FQ227136.1 | - | 2 - 3 | 159 | 66 | ||
FQ227567.1 | - | 2 - 3 | 150 | 66 | ||
FQ227951.1 | - | 2 - 3 | 150 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
