Assay Details
| Gene Symbol: Pstpip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271281.1 | NP_001258210.1 | 5 - 6 | 528 | 69 | |
| XM_008772138.1 | XP_008770360.1 | 4 - 5 | 781 | 69 | ||
| XM_008772139.1 | XP_008770361.1 | 4 - 5 | 781 | 69 | ||
| GenBank mRNA | FQ120558.1 | - | 5 - 6 | 528 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pstpip2
Genomic Map