Assay Details
| Gene Symbol: Atp5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023093.1 | NP_075581.1 | 7 - 8 | 997 | 69 | |
| GenBank mRNA | BC061830.1 | - | 7 - 8 | 997 | 5599635 | 69 |
| FQ214146.1 | - | 6 - 7 | 839 | 69 | ||
| FQ225416.1 | - | 2 - 3 | 312 | 69 | ||
| FQ225735.1 | - | 2 - 3 | 312 | 69 | ||
| FQ227531.1 | - | 2 - 3 | 312 | 69 | ||
| J05266.1 | - | 7 - 8 | 918 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp5a1
Genomic Map