Assay Details
| Gene Symbol: Atp5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023093.1 | NP_075581.1 | 3 - 4 | 353 | 68 | |
| GenBank mRNA | BC061830.1 | - | 3 - 4 | 353 | 5599635 | 68 |
| FQ214031.1 | - | 2 - 3 | 195 | 68 | ||
| FQ214146.1 | - | 2 - 3 | 195 | 68 | ||
| J05266.1 | - | 3 - 4 | 274 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp5a1
Genomic Map