Assay Details
| Gene Symbol: Naca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105939.2 | NP_001099409.1 | 4 - 5 | 339 | 71 | |
| NM_001198562.1 | NP_001185491.1 | 4 - 5 | 301 | 71 | ||
| NM_001198580.1 | NP_001185509.1 | 4 - 5 | 6079 | 71 | ||
| GenBank mRNA | BC166934.1 | - | 4 - 5 | 289 | 6922001 | 71 |
| BP494723.1 | - | 4 - 5 | 323 | 71 | ||
| FQ221678.1 | - | 4 - 5 | 334 | 71 | ||
| FQ222263.1 | - | 4 - 5 | 274 | 71 | ||
| FQ231879.1 | - | 4 - 5 | 328 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Naca
Genomic Map