Assay Details
| Gene Symbol: Sarnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033070.1 | NP_001028242.1 | 6 - 7 | 537 | 93 | |
| XM_006240781.2 | XP_006240843.1 | 6 - 7 | 566 | 93 | ||
| XM_008765031.1 | XP_008763253.1 | 6 - 7 | 561 | 93 | ||
| GenBank mRNA | BC100070.1 | - | 6 - 7 | 537 | 7385206 | 93 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sarnp
Genomic Map