Assay Details
| Gene Symbol: LOC103694201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772077.1 | XP_008770299.1 | 2 - 3 | 176 | 92 | |
| Gene Symbol: LOC100911241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749196.2 | XP_003749244.1 | 2 - 3 | 167 | 92 | |
| Gene Symbol: Spink12l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001061351.3 | XP_001061351.1 | 2 - 3 | 167 | 92 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103694201
Genomic Map