Assay Details
| Gene Symbol: MGC94891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044277.1 | NP_001037742.2 | 4 - 5 | 937 | 85 | |
| GenBank mRNA | BC083819.1 | - | 4 - 5 | 937 | 7111103 | 85 |
More Information
Updated Mapping Information
Set Membership: |
|

MGC94891
Genomic Map