Assay Details
Gene Symbol: Crb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025661.1 | NP_001020832.1 | 1 - 2 | 56 | 84 | |
GenBank mRNA | BC097382.1 | - | 1 - 2 | 56 | 7446098 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC097382.1, NM_001025661.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
