Assay Details
| Gene Symbol: Crb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025661.1 | NP_001020832.1 | 2 - 3 | 227 | 76 | |
| XM_006244303.2 | XP_006244365.1 | 2 - 3 | 1344 | 76 | ||
| GenBank mRNA | BC097382.1 | - | 2 - 3 | 227 | 7446098 | 76 |
More Information
Updated Mapping Information
Set Membership: |
|

Crb3
Genomic Map