Assay Details
| Gene Symbol: Crb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244303.2 | XP_006244365.1 | 1 - 2 | 1171 | 84 | |
| GenBank mRNA | FQ223867.1 | - | 287 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006244303.2 transcripts.
Set Membership: |
|

Crb3
Genomic Map