Assay Details
| Gene Symbol: Tnfaip8l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244371.2 | XP_006244433.1 | 1 - 2 | 149 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006244371.2 transcripts.
Set Membership: |
|

Tnfaip8l1
Genomic Map