Assay Details
| Gene Symbol: Hprt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012583.2 | NP_036715.1 | 4 - 5 | 451 | 100 | |
| GenBank mRNA | BC098629.1 | - | 4 - 5 | 451 | 7386548 | 100 |
| M63983.1 | - | 4 - 5 | 483 | 100 | ||
| M86443.1 | - | 211 | 100 | |||
| S79292.1 | - | 4 - 5 | 483 | 100 | ||
| X62085.1 | - | 411 | 100 | |||
| Gene Symbol: LOC103689983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773659.1 | XP_008771881.1 | 4 - 5 | 527 | 100 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hprt1
Genomic Map