Assay Details
Gene Symbol: Aaed1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109114.1 | NP_001102584.1 | 4 - 5 | 469 | 169 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P075421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482752 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31387 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P075421 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P075421 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P075421 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P377515 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 1695 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10793468 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793470 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793469 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793471 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10793467 | - | - |
Rat Genome 230 2.0 Array | 1382170_PM_at | - | - |
Rat Genome 230 2.0 Array | 1383443_PM_at | - | - |
Rat Genome 230 2.0 Array | 1382170_at | - | - |
Rat Genome 230 2.0 Array | 1383443_at | - | - |
Affymetrix HT Rat Focus Array | 1382170_at | - | - |
Affymetrix HT Rat Focus Array | 1383443_at | - | - |
Affymetrix HT Rat Focus Array | 1383443_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1382170_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1700008202.rn.1 | TC1700001187.rn.1 | RTA TC |
Rat Expression Set 230 | 1382170_PM_at | - | - |
Rat Expression Set 230 | 1383443_PM_at | - | - |
Rat Expression Set 230 | 1383443_at | - | - |
Rat Expression Set 230 | 1382170_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793467 | - | - |
Rat Genome U34 Set | rc_AA799427_at | - | - |
Rat Genome U34 Set | rc_AI146079_at | - | - |
