Assay Details
| Gene Symbol: Emr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007558.1 | NP_001007559.1 | 1555 | 166 | ||
| XM_008766767.1 | XP_008764989.1 | 1543 | 166 | |||
| GenBank mRNA | AY686633.1 | - | 1555 | 166 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Emr4
Genomic Map