Assay Details
Gene Symbol: LOC102553065 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257725.2 | XP_006257787.2 | 4 - 5 | 455 | 56 |
Gene Symbol: Stag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053730.1 | NP_446182.1 | 20 - 21 | 2221 | 56 | |
XM_006249020.2 | XP_006249082.1 | 20 - 21 | 2389 | 56 | ||
GenBank mRNA | AY027880.1 | - | 20 - 21 | 2221 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
