Assay Details
Gene Symbol: LOC102553065 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257725.2 | XP_006257787.2 | 6 - 7 | 630 | 90 |
Gene Symbol: Stag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053730.1 | NP_446182.1 | 22 - 23 | 2396 | 90 | |
XM_006249020.2 | XP_006249082.1 | 22 - 23 | 2564 | 90 | ||
GenBank mRNA | AY027880.1 | - | 22 - 23 | 2396 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
