Assay Details
| Gene Symbol: LOC102553065 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006257725.2 | XP_006257787.2 | 8 - 9 | 890 | 87 | |
| Gene Symbol: Stag3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053730.1 | NP_446182.1 | 24 - 25 | 2656 | 87 | |
| XM_006249020.2 | XP_006249082.1 | 24 - 25 | 2824 | 87 | ||
| GenBank mRNA | AY027880.1 | - | 24 - 25 | 2656 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC102553065
Genomic Map