Assay Details
Gene Symbol: LOC102553065 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257725.2 | XP_006257787.2 | 9 - 10 | 1029 | 87 |
Gene Symbol: Stag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053730.1 | NP_446182.1 | 25 - 26 | 2795 | 87 | |
XM_006249020.2 | XP_006249082.1 | 25 - 26 | 2963 | 87 | ||
GenBank mRNA | AY027880.1 | - | 25 - 26 | 2795 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
