Assay Details
Gene Symbol: RGD1305455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024969.2 | NP_001020140.2 | 3 - 4 | 801 | 54 | |
GenBank mRNA | DY317491.1 | - | 169 | 54 | ||
FQ211718.1 | - | 3 - 4 | 847 | 54 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
