Assay Details
| Gene Symbol: RGD1305455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024969.2 | NP_001020140.2 | 8 - 9 | 1402 | 77 | |
| GenBank mRNA | CO394313.1 | - | 3 - 4 | 347 | 77 | |
| FQ211718.1 | - | 8 - 9 | 1448 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

RGD1305455
Genomic Map