Assay Details
Gene Symbol: Wash | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127390.1 | NP_001120862.1 | 343 | 75 | ||
GenBank mRNA | BC166761.1 | - | 343 | 7930951 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
